siRNA Details
| Gene Symbol: 4933406K04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015512.2 | 1 | 94 | |||
| GenBank mRNA | AK015208.1 | - | 1 | 95 | ||
| AK015628.1 | - | 1 | 104 | |||
| AK016698.1 | - | 1 | 134 | |||
| AK016950.1 | - | 1 | 95 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933406K04Rik| Gene Symbol: 4933406K04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015512.2 | 1 | 94 | |||
| GenBank mRNA | AK015208.1 | - | 1 | 95 | ||
| AK015628.1 | - | 1 | 104 | |||
| AK016698.1 | - | 1 | 134 | |||
| AK016950.1 | - | 1 | 95 | |||