siRNA Details
| Gene Symbol: 4933406K04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015512.2 | 5, 6 | 653 | |||
| GenBank mRNA | AK015208.1 | - | 5, 6 | 654 | ||
| AK015628.1 | - | 5, 6 | 663 | |||
| AK016698.1 | - | 5, 6 | 692 | |||
| AK016950.1 | - | 4, 5 | 548 | |||

4933406K04Rik
Genomic Map