siRNA Details
| Gene Symbol: Gm12238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028480.1 | 1 | 48 | |||
| GenBank mRNA | AF357392.1 | - | 1 | 48 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12238| Gene Symbol: Gm12238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028480.1 | 1 | 48 | |||
| GenBank mRNA | AF357392.1 | - | 1 | 48 | ||