siRNA Details
| Gene Symbol: 9430076C15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015553.2 | 5 | 1620 | |||
| GenBank mRNA | AK051275.1 | - | 5 | 1620 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9430076C15Rik| Gene Symbol: 9430076C15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015553.2 | 5 | 1620 | |||
| GenBank mRNA | AK051275.1 | - | 5 | 1620 | ||