siRNA Details
| Gene Symbol: 9430076C15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015553.2 | 1, 2 | 75 | |||
| GenBank mRNA | AK034642.1 | - | 50 | |||
| AK035031.1 | - | 1, 2 | 101 | |||
| AK051275.1 | - | 1, 2 | 75 | |||
| AK166503.1 | - | 1, 2 | 87 | |||
| Gene Symbol: Creb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK170766.1 | - | 55 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB656461.1 | - | 1, 2 | 75 | Chr. 6 - 53287295 - 53394843 [+] on Build GRCm38 | |

9430076C15Rik
Genomic Map