siRNA Details
| Gene Symbol: 4933406I18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029437.1 | 1 | 192 | |||
| NR_029438.1 | 1 | 192 | ||||
| GenBank mRNA | AK016630.1 | - | 1 | 53 | ||
| AK016693.1 | - | 1 | 198 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933406I18Rik| Gene Symbol: 4933406I18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_029437.1 | 1 | 192 | |||
| NR_029438.1 | 1 | 192 | ||||
| GenBank mRNA | AK016630.1 | - | 1 | 53 | ||
| AK016693.1 | - | 1 | 198 | |||