siRNA Details
| Gene Symbol: 2900092D14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027891.1 | 1 | 674 | |||
| GenBank mRNA | AK013838.1 | - | 1 | 675 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU610322.1 | - | 1 | 412 | Chr. 1 - 42700597 - 42701253 [+] on Build GRCm38 | |

2900092D14Rik
Genomic Map