siRNA Details
| Gene Symbol: 2900092D14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027891.1 | 1 | 2722 | |||
| GenBank mRNA | AK038597.1 | - | 1 | 1913 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB174009.2 | - | 1 | 605 | Chr. 1 - 42702517 - 42703176 [+] on Build GRCm38 | |

2900092D14Rik
Genomic Map