siRNA Details
| Gene Symbol: 2900092D14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027891.1 | 1 | 842 | |||
| GenBank mRNA | AK013838.1 | - | 1 | 844 | ||
| AK038597.1 | - | 1 | 29 | |||
| AK053590.1 | - | 1 | 22 | |||
| AK163519.1 | - | 1 | 28 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2900092D14Rik| Gene Symbol: 2900092D14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027891.1 | 1 | 842 | |||
| GenBank mRNA | AK013838.1 | - | 1 | 844 | ||
| AK038597.1 | - | 1 | 29 | |||
| AK053590.1 | - | 1 | 22 | |||
| AK163519.1 | - | 1 | 28 | |||