siRNA Details
| Gene Symbol: Rnu6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | M10329.1 | - | 44 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnu6| Gene Symbol: Rnu6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | M10329.1 | - | 44 | |||