siRNA Details
| Gene Symbol: Trappc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003546.1 | 2 | 1332 | |||
| GenBank mRNA | AK012141.1 | - | 2 | 1307 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trappc13| Gene Symbol: Trappc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003546.1 | 2 | 1332 | |||
| GenBank mRNA | AK012141.1 | - | 2 | 1307 | ||