siRNA Details
| Gene Symbol: Trappc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003546.1 | 2 | 478 | |||
| GenBank mRNA | AK012141.1 | - | 2 | 453 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CB233627.1 | - | 2 | 433 | Chr. 13 - 104174575 - 104178418 [-] on Build GRCm38 | |

Trappc13
Genomic Map