siRNA Details
| Gene Symbol: Krit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033173.1 | 1, 2 | 411 | |||
| GenBank mRNA | BC054819.1 | - | 1, 2 | 403 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BQ900689.1 | - | 1, 2 | 421 | Chr. 5 - 3803182 - 3809733 [+] on Build GRCm38 | |

Krit1
Genomic Map