siRNA Details
| Gene Symbol: Krit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033173.1 | 9 | 1489 | |||
| GenBank mRNA | BC044820.1 | - | 8 | 1131 | ||
| BC054819.1 | - | 9 | 1481 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krit1| Gene Symbol: Krit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033173.1 | 9 | 1489 | |||
| GenBank mRNA | BC044820.1 | - | 8 | 1131 | ||
| BC054819.1 | - | 9 | 1481 | |||