siRNA Details
Gene Symbol: Gm3428 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030730.1 | 4 | 476 | |||
GenBank mRNA | AK136639.1 | - | 4 | 479 | ||
BY721010.1 | - | 4 | 475 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm3428 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_030730.1 | 4 | 476 | |||
GenBank mRNA | AK136639.1 | - | 4 | 479 | ||
BY721010.1 | - | 4 | 475 |