siRNA Details
| Gene Symbol: Mx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003520.1 | 9 | 2653 | |||
| GenBank mRNA | AK076430.1 | - | 14 | 2924 | ||
| AK150023.1 | - | 9 | 2652 | |||
| AK150236.1 | - | 9 | 2652 | |||
| M21039.1 | - | 11 | 2748 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mx1| Gene Symbol: Mx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003520.1 | 9 | 2653 | |||
| GenBank mRNA | AK076430.1 | - | 14 | 2924 | ||
| AK150023.1 | - | 9 | 2652 | |||
| AK150236.1 | - | 9 | 2652 | |||
| M21039.1 | - | 11 | 2748 | |||