siRNA Details
| Gene Symbol: Mx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003508.1 | 8 | 1096 | |||
| GenBank mRNA | AK143831.1 | - | 8 | 1097 | ||
| AK147949.1 | - | 1716 | ||||
| BC007127.1 | - | 8 | 1079 | 3591798 | ||
| J03368.1 | - | 8 | 1094 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mx2| Gene Symbol: Mx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003508.1 | 8 | 1096 | |||
| GenBank mRNA | AK143831.1 | - | 8 | 1097 | ||
| AK147949.1 | - | 1716 | ||||
| BC007127.1 | - | 8 | 1079 | 3591798 | ||
| J03368.1 | - | 8 | 1094 | |||