siRNA Details
| Gene Symbol: 4831440E17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030700.1 | 1 | 440 | |||
| GenBank mRNA | AK029244.1 | - | 1 | 296 | ||
| AK149324.1 | - | 1 | 439 | |||
| AK163107.1 | - | 1 | 67 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4831440E17Rik| Gene Symbol: 4831440E17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030700.1 | 1 | 440 | |||
| GenBank mRNA | AK029244.1 | - | 1 | 296 | ||
| AK149324.1 | - | 1 | 439 | |||
| AK163107.1 | - | 1 | 67 | |||