siRNA Details
| Gene Symbol: 9530036O11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015562.1 | 4 | 2140 | |||
| GenBank mRNA | AK035474.1 | - | 4 | 2140 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9530036O11Rik| Gene Symbol: 9530036O11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015562.1 | 4 | 2140 | |||
| GenBank mRNA | AK035474.1 | - | 4 | 2140 | ||