siRNA Details
| Gene Symbol: Gm10584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028578.1 | 1 | 1710 | |||
| GenBank mRNA | AK042978.1 | - | 1 | 929 | ||
| AK141912.1 | - | 1 | 1710 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm10584| Gene Symbol: Gm10584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028578.1 | 1 | 1710 | |||
| GenBank mRNA | AK042978.1 | - | 1 | 929 | ||
| AK141912.1 | - | 1 | 1710 | |||