siRNA Details
| Gene Symbol: F930015N05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028445.1 | 1 | 3194 | |||
| GenBank mRNA | AA797660.1 | - | 1 | 140 | ||
| AK158006.1 | - | 1 | 3026 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F930015N05Rik| Gene Symbol: F930015N05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028445.1 | 1 | 3194 | |||
| GenBank mRNA | AA797660.1 | - | 1 | 140 | ||
| AK158006.1 | - | 1 | 3026 | |||