siRNA Details
| Gene Symbol: Psg-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002857.1 | 3 | 856 | |||
| GenBank mRNA | AK132281.1 | - | 3 | 856 | ||
| M83348.1 | - | 3 | 852 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CF551674.1 | - | 3 | 906 | Chr. 7 - 17672292 - 17678005 [+] on Build GRCm38 | |

Psg-ps1
Genomic Map