siRNA Details
| Gene Symbol: Psg-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002857.1 | 3 | 757 | |||
| GenBank mRNA | AK132281.1 | - | 3 | 757 | ||
| M83348.1 | - | 3 | 753 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psg-ps1| Gene Symbol: Psg-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002857.1 | 3 | 757 | |||
| GenBank mRNA | AK132281.1 | - | 3 | 757 | ||
| M83348.1 | - | 3 | 753 | |||