siRNA Details
| Gene Symbol: 1700020N18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026924.1 | 3 | 528 | |||
| GenBank mRNA | AK006182.1 | - | 3 | 529 | ||
| DQ688551.1 | - | 1 | 20 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706279.1 | - | 3 | 529 | Chr. 1 - 91404877 - 91406029 [-] on Build GRCm38 | |

1700020N18Rik
Genomic Map