siRNA Details
| Gene Symbol: 1700020N18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026924.1 | 3 | 298 | |||
| GenBank mRNA | AK006182.1 | - | 3 | 299 | ||
| AK006756.1 | - | 3 | 299 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700020N18Rik| Gene Symbol: 1700020N18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026924.1 | 3 | 298 | |||
| GenBank mRNA | AK006182.1 | - | 3 | 299 | ||
| AK006756.1 | - | 3 | 299 | |||