siRNA Details
| Gene Symbol: D930048N14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027958.1 | 4 | 1461 | |||
| GenBank mRNA | AK086740.1 | - | 4 | 1461 | ||
| AK219496.1 | - | 1 | 384 | |||
| BC075670.1 | - | 4 | 1443 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D930048N14Rik| Gene Symbol: D930048N14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027958.1 | 4 | 1461 | |||
| GenBank mRNA | AK086740.1 | - | 4 | 1461 | ||
| AK219496.1 | - | 1 | 384 | |||
| BC075670.1 | - | 4 | 1443 | |||