siRNA Details
| Gene Symbol: Yaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028315.1 | 3 | 337 | |||
| GenBank mRNA | AB100453.1 | - | 3 | 327 | ||
| AK011702.1 | - | 3 | 338 | |||
| AK012787.1 | - | 2 | 173 | |||
| AK184096.1 | - | 2 | 52 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Yaf2| Gene Symbol: Yaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028315.1 | 3 | 337 | |||
| GenBank mRNA | AB100453.1 | - | 3 | 327 | ||
| AK011702.1 | - | 3 | 338 | |||
| AK012787.1 | - | 2 | 173 | |||
| AK184096.1 | - | 2 | 52 | |||