siRNA Details
| Gene Symbol: Gm13375 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033225.1 | 2 | 1238 | |||
| GenBank mRNA | AK158585.1 | - | 1 | 1114 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13375| Gene Symbol: Gm13375 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033225.1 | 2 | 1238 | |||
| GenBank mRNA | AK158585.1 | - | 1 | 1114 | ||