siRNA Details
Gene Symbol: Gm13375 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033225.1 | 2 | 1164 | |||
GenBank mRNA | AK158585.1 | - | 1 | 1040 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm13375 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033225.1 | 2 | 1164 | |||
GenBank mRNA | AK158585.1 | - | 1 | 1040 |