siRNA Details
| Gene Symbol: 2010016I18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033207.1 | 2 | 1336 | |||
| GenBank mRNA | AK040034.1 | - | 1 | 1247 | ||
| AK080971.1 | - | 2 | 1337 | |||
| AK140363.1 | - | 2 | 1337 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2010016I18Rik| Gene Symbol: 2010016I18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033207.1 | 2 | 1336 | |||
| GenBank mRNA | AK040034.1 | - | 1 | 1247 | ||
| AK080971.1 | - | 2 | 1337 | |||
| AK140363.1 | - | 2 | 1337 | |||