siRNA Details
| Gene Symbol: A630072M18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030699.1 | 1 | 1277 | |||
| GenBank mRNA | AK040438.1 | - | 1 | 283 | ||
| AK042225.1 | - | 1 | 292 | |||
| AK086970.1 | - | 1 | 277 | |||
| AK163124.1 | - | 1 | 1277 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A630072M18Rik| Gene Symbol: A630072M18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_030699.1 | 1 | 1277 | |||
| GenBank mRNA | AK040438.1 | - | 1 | 283 | ||
| AK042225.1 | - | 1 | 292 | |||
| AK086970.1 | - | 1 | 277 | |||
| AK163124.1 | - | 1 | 1277 | |||