siRNA Details
| Gene Symbol: Zfp862-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015597.1 | 3 | 990 | |||
| NR_027808.1 | 3 | 990 | ||||
| GenBank mRNA | AK129160.1 | - | 3 | 912 | ||
| BC068148.1 | - | 3 | 941 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp862-ps| Gene Symbol: Zfp862-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015597.1 | 3 | 990 | |||
| NR_027808.1 | 3 | 990 | ||||
| GenBank mRNA | AK129160.1 | - | 3 | 912 | ||
| BC068148.1 | - | 3 | 941 | |||