siRNA Details
| Gene Symbol: Zfp862-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015597.1 | 7 | 3995 | |||
| NR_027808.1 | 7 | 3919 | ||||
| GenBank mRNA | AK028840.1 | - | 2 | 1351 | ||
| AK129160.1 | - | 7 | 3919 | |||
| BC037484.1 | - | 1 | 187 | |||
| BC059055.1 | - | 2 | 1968 | |||
| BC068148.1 | - | 7 | 3870 | |||

Zfp862-ps
Genomic Map