siRNA Details
| Gene Symbol: Snord118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028566.1 | 1 | 50 | |||
| GenBank mRNA | AF357360.1 | - | 1 | 39 | ||
| X04239.2 | - | 1 | 50 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snord118| Gene Symbol: Snord118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028566.1 | 1 | 50 | |||
| GenBank mRNA | AF357360.1 | - | 1 | 39 | ||
| X04239.2 | - | 1 | 50 | |||