siRNA Details
| Gene Symbol: Vash2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027352.1 | 1 | 90 | |||
| GenBank mRNA | AK045261.1 | - | 1 | 19 | ||
| AK078596.1 | - | 1 | 91 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vash2| Gene Symbol: Vash2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027352.1 | 1 | 90 | |||
| GenBank mRNA | AK045261.1 | - | 1 | 19 | ||
| AK078596.1 | - | 1 | 91 | |||