siRNA Details
| Gene Symbol: Ngrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028053.1 | 1 | 540 | |||
| GenBank mRNA | AK083330.1 | - | 1 | 536 | ||
| AK146059.1 | - | 1 | 518 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ngrn| Gene Symbol: Ngrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028053.1 | 1 | 540 | |||
| GenBank mRNA | AK083330.1 | - | 1 | 536 | ||
| AK146059.1 | - | 1 | 518 | |||