siRNA Details
| Gene Symbol: Zfp809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028420.1 | 3 | 297 | |||
| GenBank mRNA | AK089532.1 | - | 3 | 278 | ||
| BB658893.1 | - | 3 | 278 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp809| Gene Symbol: Zfp809 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028420.1 | 3 | 297 | |||
| GenBank mRNA | AK089532.1 | - | 3 | 278 | ||
| BB658893.1 | - | 3 | 278 | |||