siRNA Details
| Gene Symbol: Speer6-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001581.2 | 2, 3 | 218 | |||
| GenBank mRNA | AK006638.1 | - | 2, 3 | 219 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Speer6-ps1| Gene Symbol: Speer6-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001581.2 | 2, 3 | 218 | |||
| GenBank mRNA | AK006638.1 | - | 2, 3 | 219 | ||