siRNA Details
| Gene Symbol: Taf1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028401.1 | 8 | 1045 | |||
| GenBank mRNA | AK012656.1 | - | 6 | 894 | ||
| AK014920.1 | - | 8 | 997 | |||
| AK020036.1 | - | 7 | 957 | |||
| AK050165.1 | - | 8 | 1001 | |||
| AK161656.1 | - | 7 | 2153 | |||
| AK166329.1 | - | 8 | 1000 | |||
| AK166633.1 | - | 7 | 2115 | |||
| BC056964.1 | - | 5 | 3216 | |||
| BC110660.1 | - | 7 | 987 | 3158156 | ||

Taf1d
Genomic Map