siRNA Details
| Gene Symbol: 1700108F19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015485.1 | 5 | 774 | |||
| GenBank mRNA | AK006165.1 | - | 4 | 1334 | ||
| AK007133.1 | - | 5 | 764 | |||
| BY707214.1 | - | 5 | 762 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700108F19Rik| Gene Symbol: 1700108F19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015485.1 | 5 | 774 | |||
| GenBank mRNA | AK006165.1 | - | 4 | 1334 | ||
| AK007133.1 | - | 5 | 764 | |||
| BY707214.1 | - | 5 | 762 | |||