siRNA Details
| Gene Symbol: 1700108F19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015485.1 | 5 | 659 | |||
| GenBank mRNA | AK006165.1 | - | 4 | 1219 | ||
| AK007133.1 | - | 5 | 649 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CN841323.1 | - | 4 | 429 | Chr. 14 - 76677607 - 76687107 [+] on Build GRCm38 | |

1700108F19Rik
Genomic Map