siRNA Details
| Gene Symbol: 1700108F19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015485.1 | 2, 3 | 247 | |||
| GenBank mRNA | AK006165.1 | - | 1, 2 | 807 | ||
| AK007133.1 | - | 2, 3 | 237 | |||
| AK076702.1 | - | 2, 3 | 206 | |||
| BY707214.1 | - | 2, 3 | 235 | |||

1700108F19Rik
Genomic Map