siRNA Details
Gene Symbol: Daw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027636.1 | 5, 6 | 454 | |||
GenBank mRNA | AK016201.1 | - | 5, 6 | 449 | ||
BU612798.1 | - | 5, 6 | 417 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Daw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027636.1 | 5, 6 | 454 | |||
GenBank mRNA | AK016201.1 | - | 5, 6 | 449 | ||
BU612798.1 | - | 5, 6 | 417 |