siRNA Details
| Gene Symbol: Daw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027636.1 | 2, 3 | 147 | |||
| GenBank mRNA | AK016201.1 | - | 2, 3 | 141 | ||
| BU612798.1 | - | 2, 3 | 110 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Daw1| Gene Symbol: Daw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027636.1 | 2, 3 | 147 | |||
| GenBank mRNA | AK016201.1 | - | 2, 3 | 141 | ||
| BU612798.1 | - | 2, 3 | 110 | |||