siRNA Details
| Gene Symbol: 1700031M16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015496.1 | 10, 11 | 774 | |||
| GenBank mRNA | BC048654.1 | - | 10, 11 | 774 | ||
| BC099572.1 | - | 10, 11 | 779 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700031M16Rik| Gene Symbol: 1700031M16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015496.1 | 10, 11 | 774 | |||
| GenBank mRNA | BC048654.1 | - | 10, 11 | 774 | ||
| BC099572.1 | - | 10, 11 | 779 | |||