siRNA Details
| Gene Symbol: Btnl5-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_004051.1 | 9 | 2567 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btnl5-ps| Gene Symbol: Btnl5-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_004051.1 | 9 | 2567 | |||