siRNA Details
| Gene Symbol: 2610306M01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028298.1 | 1 | 359 | |||
| GenBank mRNA | AK011997.1 | - | 1 | 360 | ||
| AK015614.1 | - | 1 | 334 | |||
| BC058130.1 | - | 1 | 72 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610306M01Rik| Gene Symbol: 2610306M01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028298.1 | 1 | 359 | |||
| GenBank mRNA | AK011997.1 | - | 1 | 360 | ||
| AK015614.1 | - | 1 | 334 | |||
| BC058130.1 | - | 1 | 72 | |||