siRNA Details
Gene Symbol: Ube2d2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005969.1 | - | 1 | 42 | ||
AK133385.1 | - | 1 | 24 | |||
BC050749.1 | - | 1 | 41 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ube2d2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005969.1 | - | 1 | 42 | ||
AK133385.1 | - | 1 | 24 | |||
BC050749.1 | - | 1 | 41 |