siRNA Details
| Gene Symbol: Ube2d2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005969.1 | - | 1 | 671 | ||
| AK133385.1 | - | 1 | 653 | |||
| BC050749.1 | - | 1 | 670 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2d2b| Gene Symbol: Ube2d2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005969.1 | - | 1 | 671 | ||
| AK133385.1 | - | 1 | 653 | |||
| BC050749.1 | - | 1 | 670 | |||