siRNA Details
| Gene Symbol: Gm7367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003376.2 | 1 | 14 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm7367| Gene Symbol: Gm7367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003376.2 | 1 | 14 | |||